Gene Transcripts

Gene Symbol
Thada
Entrez Gene ID
240174
Gene Name
thyroid adenoma associated
Gene Aliases
6530405K19, A930021O22, AU019899, BC052885, GITA
Chromosome Location
Chr. 17: 84190055 - 84466261 on Build GRCm38
UniGene ID
Mm.246767
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183021.3 NP_898842.2 9 927
GenBank mRNA AK044550.1 - 9 880
AK154078.1 - 9 925
EF222207.1 - 9 835

View Full Product Details