Gene Transcripts

Gene Symbol
Gin1
Entrez Gene ID
252876
Gene Name
gypsy retrotransposon integrase 1
Gene Aliases
4930429M06Rik, 8430411H10Rik, Gin-1, Zh2c2
Chromosome Location
Chr. 1: 97770168 - 97793710 on Build GRCm38
UniGene ID
Mm.86090
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026250.2 NP_080526.2 4 537
GenBank mRNA AK014287.1 - 4 549
AK015243.1 - 4 516
AK169361.1 - 4 500
BC033572.1 - 3 482 5067692

View Full Product Details