Gene Transcripts

Gene Symbol
Olfr76
Entrez Gene ID
258677
Gene Name
olfactory receptor 76
Gene Aliases
MOR215-3, V5
Chromosome Location
Chr. 19: 12119721 - 12120674 on Build GRCm38
UniGene ID
Mm.261576
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146682.1 NP_666893.1 1 573
GenBank mRNA BC138072.1 - 1 599
BC138073.1 - 1 599 8861092
Y15525.1 - 1 387

View Full Product Details