Gene Transcripts

Gene Symbol
Wfdc16
Entrez Gene ID
277345
Gene Name
WAP four-disulfide core domain 16
Gene Aliases
BC043944, WAP16
Chromosome Location
Chr. 2: 164634701 - 164638808 on Build GRCm38
UniGene ID
Mm.247244
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012723.2 NP_001012741.2 4 926
GenBank mRNA AY542488.1 - 5 512
AY542489.1 - 5 510
BC043944.1 - 4 912

View Full Product Details