Gene Transcripts

Gene Symbol
Lrrc58
Entrez Gene ID
320184
Gene Name
leucine rich repeat containing 58
Gene Aliases
1810012N18Rik, C330018J07Rik
Chromosome Location
Chr. 16: 37868400 - 37888857 on Build GRCm38
UniGene ID
Mm.390882
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177093.3 NP_796067.2 3 937
GenBank mRNA AK049274.1 - 3 938
AK090204.1 - 3 946
AK147818.1 - 3 913
BC141007.1 - 3 897 9055619

View Full Product Details