Gene Transcripts

Gene Symbol
Vmn2r12
Entrez Gene ID
627569
Gene Name
vomeronasal 2, receptor 12
Gene Aliases
EG627569, Gm6769
Chromosome Location
Chr. 5: 109085849 - 109097864 on Build GRCm38
UniGene ID
Mm.469884
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104623.1 NP_001098093.1 6 2175

Gene Symbol
Vmn2r11
Entrez Gene ID
384219
Gene Name
vomeronasal 2, receptor 11
Gene Aliases
EG384219
Chromosome Location
Chr. 5: 109046873 - 109059452 on Build GRCm38
UniGene ID
Mm.469885
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104622.1 NP_001098092.1 6 2220

Gene Symbol
Vmn2r8
Entrez Gene ID
627479
Gene Name
vomeronasal 2, receptor 8
Gene Aliases
EG627479
Chromosome Location
Chr. 5: 108797193 - 108808754 on Build GRCm38
UniGene ID
Mm.469886
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104620.1 NP_001098090.1 6 2181

Gene Symbol
Vmn2r9
Entrez Gene ID
435864
Gene Name
vomeronasal 2, receptor 9
Gene Aliases
EG435864
Chromosome Location
Chr. 5: 108842947 - 108852510 on Build GRCm38
UniGene ID
Mm.387745
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104621.1 NP_001098091.1 6 2181

View Full Product Details