Gene Transcripts

Gene Symbol
RPL3L
Entrez Gene ID
6123
Gene Name
ribosomal protein L3 like
Gene Aliases
Chromosome Location
Chr. 16: 1944571 - 1955123 on Build GRCh38
UniGene ID
Hs.657266
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005061.2 NP_005052.1 8 1083
XM_011522571.2 XP_011520873.1 8 1543
GenBank mRNA AK313162.1 - 8 1095
BC050413.1 - 8 1075 6204847
U65581.1 - 8 1084

View Full Product Details