Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013777.2 | NP_038805.2 | 4 | 454 | |
GenBank mRNA | AB027237.1 | - | 4 | 424 | |
AF177041.1 | - | 4 | 445 | ||
AK007716.1 | - | 4 | 452 | ||
BC012643.1 | - | 4 | 428 | 4205715 | |
BC063780.1 | - | 4 | 431 | 4219625 |