Gene Transcripts

Gene Symbol
Slc38a6
Entrez Gene ID
625098
Gene Name
solute carrier family 38, member 6
Gene Aliases
AW322671, EG625098
Chromosome Location
Chr. 12: 73286794 - 73361998 on Build GRCm38
UniGene ID
Mm.32075
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037717.3 NP_001032806.2 14 1148
GenBank mRNA BC043932.1 - 7 475
BC157917.1 - 14 1097 9007345
BC157928.1 - 14 1097 9088101

View Full Product Details