Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252089.1 | NP_001239018.1 | 2 | 455 | |
NM_009033.2 | NP_033059.2 | 2 | 501 | ||
GenBank mRNA | AF031568.1 | - | 2 | 367 | |
AK041927.1 | - | 2 | 392 | ||
AK088213.1 | - | 2 | 388 | ||
AK149658.1 | - | 2 | 474 | ||
AK149764.1 | - | 2 | 474 | ||
AK218950.1 | - | 1 | 251 | ||
BC011441.1 | - | 2 | 406 | 3153831 | |
BC089350.1 | - | 2 | 415 | 30359767 |