Gene Transcripts

Gene Symbol
ATXN1
Entrez Gene ID
6310
Gene Name
ataxin 1
Gene Aliases
ATX1, D6S504E, SCA1
Chromosome Location
Chr. 6: 16299112 - 16761490 on Build GRCh38
UniGene ID
Hs.434961
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000332.3 NP_000323.2 8 2793
NM_001128164.1 NP_001121636.1 7 2744
GenBank mRNA BC117125.1 - 1 1855
X79204.1 - 8 2760

View Full Product Details