Gene Transcripts
- Gene Symbol
- Rhox5
- Entrez Gene ID
- 24631
- Gene Name
- reproductive homeobox 5
- Gene Aliases
- Pem
- Chromosome Location
- Chr. X: 123900363 - 123916988 on Build Rnor_6.0
- UniGene ID
- Rn.44483
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022175.1 |
NP_071511.1
|
3, 4 |
492 |
|
GenBank mRNA |
DQ058653.1 |
- |
3, 4 |
492 |
|
View Full Product Details