Gene Transcripts

Gene Symbol
Cym
Entrez Gene ID
56825
Gene Name
chymosin
Gene Aliases
Chromosome Location
Chr. 2: 209962848 - 209973205 on Build Rnor_6.0
UniGene ID
Rn.48676
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020091.1 NP_064476.1 3 249
GenBank mRNA AJ251688.1 - 3 249

View Full Product Details