Gene Transcripts
- Gene Symbol
- SPP2
- Entrez Gene ID
- 6694
- Gene Name
- secreted phosphoprotein 2
- Gene Aliases
- SPP-24, SPP24
- Chromosome Location
- Chr. 2: 234050671 - 234077134 on Build GRCh38
- UniGene ID
- Hs.444488
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006944.2 |
NP_008875.1
|
2, 3 |
290 |
|
XM_005246102.3 |
XP_005246159.1
|
2, 3 |
321 |
|
XM_011511698.2 |
XP_011510000.1
|
2, 3 |
321 |
|
XM_011511699.2 |
XP_011510001.1
|
2, 3 |
321 |
|
XM_011511700.2 |
XP_011510002.1
|
2, 3 |
321 |
|
GenBank mRNA |
AJ308099.1 |
- |
2, 3 |
290 |
|
BC069401.1 |
- |
2, 3 |
205 |
7262158 |
BC106705.1 |
- |
2, 3 |
205 |
|
BC112438.1 |
- |
2, 3 |
205 |
|
U20530.1 |
- |
2, 3 |
205 |
|
View Full Product Details