Gene Transcripts

Gene Symbol
SSR4
Entrez Gene ID
6748
Gene Name
signal sequence receptor subunit 4
Gene Aliases
CDG1Y, TRAPD
Chromosome Location
Chr. X: 153794175 - 153798512 on Build GRCh38
UniGene ID
Hs.409223
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204526.1 NP_001191455.1 6 544
NM_001204527.1 NP_001191456.1 6 567
NM_006280.2 NP_006271.1 5 517
NR_037927.1 7 738
XM_011531186.1 XP_011529488.1 6 426
XM_011531187.1 XP_011529489.1 6 426
XM_017029756.1 XP_016885245.1 6 1315
XM_017029757.1 XP_016885246.1 6 1050
GenBank mRNA AK057117.1 - 4 2100
AK094395.1 - 5 2220
AK290493.1 - 5 423
BC003371.1 - 5 397 2900758
BE791319.1 - 5 439
BG165852.1 - 6 551
BP202705.1 - 6 506
BT007192.1 - 5 393
CD672615.1 - 6 609
DB238237.1 - 5 516
X90583.1 - 5 423
Z69043.1 - 5 442

View Full Product Details