Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001015878.1 | NP_001015878.1 | 4 | 506 | |
NM_001015879.1 | NP_001015879.1 | 4 | 365 | ||
NM_003160.2 | NP_003151.2 | 4 | 370 | ||
GenBank mRNA | AB017332.1 | - | 4 | 511 | |
AF059681.1 | - | 4 | 566 | ||
AK301886.1 | - | 3 | 946 | ||
AY661554.1 | - | 4 | 365 | ||
AY714054.1 | - | 4 | 308 | ||
BC075064.2 | - | 3 | 259 | 30915373 | |
EU668345.1 | - | 4 | 566 |