Gene Transcripts

Gene Symbol
Agxt2
Entrez Gene ID
83784
Gene Name
alanine-glyoxylate aminotransferase 2
Gene Aliases
AGT2
Chromosome Location
Chr. 2: 60337664 - 60379144 on Build Rnor_6.0
UniGene ID
Rn.1874
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031835.2 NP_114023.1 14 1528
GenBank mRNA AB002584.1 - 14 1499
BC081765.1 - 14 1528 7106995
FQ219625.1 - 6 609

View Full Product Details