Gene Transcripts
- Gene Symbol
- Sacm1l
- Entrez Gene ID
- 116482
- Gene Name
- SAC1 suppressor of actin mutations 1-like (yeast)
- Gene Aliases
- Sac1
- Chromosome Location
- Chr. 8: 132607166 - 132663502 on Build Rnor_6.0
- UniGene ID
- Rn.161978
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053798.2 |
NP_446250.1
|
5 |
479 |
|
View Full Product Details