Gene Transcripts
- Gene Symbol
- Serbp1
- Entrez Gene ID
- 246303
- Gene Name
- Serpine1 mRNA binding protein 1
- Gene Aliases
- Pai-Rbp1, Pairbp1, Rda288
- Chromosome Location
- Chr. 4: 98027566 - 98044705 on Build Rnor_6.0
- UniGene ID
- Rn.203108
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145086.1 |
NP_659554.1
|
7, 8 |
1180 |
|
XM_006236616.1 |
XP_006236678.1
|
7, 8 |
1273 |
|
XM_006236617.1 |
XP_006236679.1
|
7, 8 |
1255 |
|
XM_006236618.1 |
XP_006236680.1
|
7, 8 |
1210 |
|
GenBank mRNA |
AF388527.1 |
- |
7, 8 |
1186 |
|
BC079337.1 |
- |
7, 8 |
1180 |
7106027 |
FQ213573.1 |
- |
7, 8 |
1269 |
|
FQ214321.1 |
- |
7, 8 |
1207 |
|
FQ223736.1 |
- |
7, 8 |
1228 |
|
FQ225988.1 |
- |
7, 8 |
922 |
|
FQ226374.1 |
- |
7, 8 |
1201 |
|
FQ226420.1 |
- |
7, 8 |
940 |
|
FQ227636.1 |
- |
7, 8 |
1184 |
|
FQ227901.1 |
- |
3, 4 |
764 |
|
FQ228026.1 |
- |
7, 8 |
1191 |
|
FQ230135.1 |
- |
6, 7 |
1117 |
|
FQ230192.1 |
- |
7, 8 |
1191 |
|
FQ230382.1 |
- |
7, 8 |
921 |
|
FQ231329.1 |
- |
7, 8 |
895 |
|
FQ231840.1 |
- |
7, 8 |
941 |
|
FQ232288.1 |
- |
7, 8 |
1183 |
|
FQ232894.1 |
- |
7, 8 |
1202 |
|
FQ233179.1 |
- |
7, 8 |
1201 |
|
FQ233457.1 |
- |
7, 8 |
1252 |
|
FQ233487.1 |
- |
7, 8 |
877 |
|
FQ233911.1 |
- |
7, 8 |
1184 |
|
FQ234284.1 |
- |
7, 8 |
1157 |
|
FQ234534.1 |
- |
7, 8 |
1184 |
|
FQ235054.1 |
- |
7, 8 |
877 |
|
U21718.1 |
- |
6, 7 |
806 |
|
View Full Product Details