Gene Transcripts

Gene Symbol
Obp2b
Entrez Gene ID
286933
Gene Name
odorant binding protein 2B
Gene Aliases
Obp2a, Obp2aa, Ry2g12
Chromosome Location
Chr. 3: 3167744 - 3181159 on Build Rnor_6.0
UniGene ID
Rn.11371
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173148.1 NP_775171.1 397
XM_008761579.1 XP_008759801.1 5 502
GenBank mRNA M76734.1 - 397

Gene Symbol
LOC103691751
Entrez Gene ID
103691751
Gene Name
odorant-binding protein 2b-like
Gene Aliases
Chromosome Location
Chr. 3: 2999712 - 3002479 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008761619.1 XP_008759841.1 403

View Full Product Details