Gene Transcripts

Gene Symbol
Cdc45
Entrez Gene ID
287961
Gene Name
cell division cycle 45
Gene Aliases
Cdc45l
Chromosome Location
Chr. 11: 86328779 - 86354101 on Build Rnor_6.0
UniGene ID
Rn.98535
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105866.2 NP_001099336.1 3 202
XM_008768866.1 XP_008767088.1 4 524
GenBank mRNA BC161946.1 - 3 202
FQ232109.1 - 3 204

View Full Product Details