Gene Transcripts

Gene Symbol
Susd4
Entrez Gene ID
289335
Gene Name
sushi domain containing 4
Gene Aliases
RGD1564043
Chromosome Location
Chr. 13: 101181664 - 101307912 on Build Rnor_6.0
UniGene ID
Rn.19527
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105982.1 NP_001099452.1 5 829
XM_006250373.2 XP_006250435.1 5 983
XM_006250375.2 XP_006250437.1 5 868
XM_008769820.1 XP_008768042.1 5 983
XM_008769821.1 XP_008768043.1 5 872
XM_008769822.1 XP_008768044.1 5 961
XM_008769823.1 XP_008768045.1 5 983
GenBank mRNA FQ212123.1 - 5 813

View Full Product Details