Gene Transcripts

Gene Symbol
Olr8
Entrez Gene ID
292563
Gene Name
olfactory receptor 8
Gene Aliases
Chromosome Location
Chr. 1: 70495403 - 70496347 on Build Rnor_6.0
UniGene ID
Rn.142001
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000113.1 NP_001000113.1 1 106

View Full Product Details