Gene Transcripts

Gene Symbol
Slx1b
Entrez Gene ID
293489
Gene Name
SLX1 structure-specific endonuclease subunit homolog B (S. cerevisiae)
Gene Aliases
Giyd1, Giyd2, LRRGT00058, RGD1311568
Chromosome Location
Chr. 1: 198114514 - 198120061 on Build Rnor_6.0
UniGene ID
Rn.30339
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009292.1 NP_001009292.1 1 188
GenBank mRNA BC087090.1 - 1 188

View Full Product Details