Gene Transcripts

Gene Symbol
Wnt8b
Entrez Gene ID
293990
Gene Name
wingless-type MMTV integration site family, member 8B
Gene Aliases
Chromosome Location
Chr. 1: 264244200 - 264264330 on Build Rnor_6.0
UniGene ID
Rn.58872
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106359.1 NP_001099829.1 5 565
XM_006231442.2 XP_006231504.1 6 721

View Full Product Details