Gene Transcripts

Gene Symbol
VPREB1
Entrez Gene ID
7441
Gene Name
pre-B lymphocyte 1
Gene Aliases
CD179a, IGI, IGVPB, VPREB
Chromosome Location
Chr. 22: 22244780 - 22245515 on Build GRCh38
UniGene ID
Hs.247979
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007128.3 NP_009059.1 1, 2 83
GenBank mRNA BC017922.1 - 1, 2 75
CR456609.1 - 1, 2 60

View Full Product Details