Gene Transcripts

Gene Symbol
Wbp5
Entrez Gene ID
680354
Gene Name
WW domain binding protein 5
Gene Aliases
RGD1560111
Chromosome Location
Chr. X: 106791333 - 106794667 on Build Rnor_6.0
UniGene ID
Rn.6930
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127502.1 NP_001120974.1 2 177
XM_006257281.2 XP_006257343.1 2 170
XM_008773438.1 XP_008771660.1 2 170
GenBank mRNA BC166897.1 - 2 177
FQ215245.1 - 2 105
FQ220910.1 - 2 159
FQ232216.1 - 2 151

View Full Product Details