Gene Transcripts

Gene Symbol
P22k15
Entrez Gene ID
296229
Gene Name
cystatin related protein 2
Gene Aliases
Crp2
Chromosome Location
Chr. 3: 143426001 - 143430075 on Build Rnor_6.0
UniGene ID
Rn.43503
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199266.2 NP_954887.1 123
GenBank mRNA BC100632.1 - 123
M58169.1 - 154

View Full Product Details