Gene Transcripts

Gene Symbol
Angpt4
Entrez Gene ID
296269
Gene Name
angiopoietin 4
Gene Aliases
Agpt4
Chromosome Location
Chr. 3: 147422095 - 147455670 on Build Rnor_6.0
UniGene ID
Rn.9586
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106526.1 NP_001099996.1 9 1558

View Full Product Details