Gene Transcripts

Gene Symbol
Camsap1
Entrez Gene ID
296580
Gene Name
calmodulin regulated spectrin-associated protein 1
Gene Aliases
RGD1565022
Chromosome Location
Chr. 3: 3373971 - 3434027 on Build Rnor_6.0
UniGene ID
Rn.203213
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168549.1 NP_001162021.1 9 1489
XM_006233672.2 XP_006233734.1 9 1457
XM_006233673.2 XP_006233735.1 8 1424
XM_006233674.2 XP_006233736.1 8 1424
XM_006233675.2 XP_006233737.1 9 1457
XM_006233677.2 XP_006233739.1 4 369

View Full Product Details