Gene Transcripts

Gene Symbol
Pnisr
Entrez Gene ID
297942
Gene Name
PNN-interacting serine/arginine-rich protein
Gene Aliases
RGD1307395, Sfrs18, Srsf18
Chromosome Location
Chr. 5: 35991068 - 36017952 on Build Rnor_6.0
UniGene ID
Rn.8700
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025274.2 NP_001020445.2 10 1279
GenBank mRNA BC091301.1 - 1 358
FQ231658.1 - 3 126

View Full Product Details