Gene Transcripts

Gene Symbol
Rdh16
Entrez Gene ID
299511
Gene Name
retinol dehydrogenase 16 (all-trans)
Gene Aliases
Rdh2, RoDHII
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.94108
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199208.1 NP_954678.1 969
GenBank mRNA BC062000.1 - 936
BC079153.1 - 3, 4 793 7128686
U33500.1 - 969

Gene Symbol
LOC100365958
Entrez Gene ID
100365958
Gene Name
Rdh2 protein-like
Gene Aliases
Chromosome Location
Chr. 7: 71065077 - 71074128 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006241512.2 XP_006241574.1 3, 4 947

View Full Product Details