Gene Transcripts

Gene Symbol
Spink8
Entrez Gene ID
301016
Gene Name
serine peptidase inhibitor, Kazal type 8
Gene Aliases
RGD1307150
Chromosome Location
Chr. 8: 117906093 - 117917821 on Build Rnor_6.0
UniGene ID
Rn.3817
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106860.1 NP_001100330.1 4, 5 373
XM_008766512.1 XP_008764734.1 5, 6 538
GenBank mRNA BC166531.1 - 3, 4 1701

View Full Product Details