Gene Transcripts

Gene Symbol
Cdcp1
Entrez Gene ID
301082
Gene Name
CUB domain containing protein 1
Gene Aliases
RGD1305578
Chromosome Location
Chr. 8: 132260007 - 132296661 on Build Rnor_6.0
UniGene ID
Rn.7729
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106869.1 NP_001100339.1 2 390
XM_006244189.2 XP_006244251.1 2 381

View Full Product Details