Gene Transcripts

Gene Symbol
Orc2
Entrez Gene ID
301430
Gene Name
origin recognition complex, subunit 2
Gene Aliases
Orc2l
Chromosome Location
Chr. 9: 65350630 - 65387278 on Build Rnor_6.0
UniGene ID
Rn.228665
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012003.1 NP_001012003.1 7 539
XM_006244956.2 XP_006245018.1 8 825
GenBank mRNA AB164704.1 - 7 539
BC129076.1 - 7 673

View Full Product Details