Gene Transcripts
- Gene Symbol
- REEP5
- Entrez Gene ID
- 7905
- Gene Name
- receptor accessory protein 5
- Gene Aliases
- C5orf18, D5S346, DP1, TB2, YOP1, Yip2e
- Chromosome Location
- Chr. 5: 112876384 - 112922334 on Build GRCh38
- UniGene ID
- Hs.429608
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005669.4 |
NP_005660.4
|
1, 2 |
263 |
|
XM_017009844.1 |
XP_016865333.1
|
1, 2 |
180 |
|
GenBank mRNA |
AK295436.1 |
- |
1, 2 |
154 |
|
AK298268.1 |
- |
1, 2 |
167 |
|
AK299902.1 |
- |
1, 2 |
159 |
|
AK300048.1 |
- |
1, 2 |
168 |
|
AK312522.1 |
- |
1, 2 |
168 |
|
AY562243.1 |
- |
1, 2 |
119 |
|
BC000232.1 |
- |
1, 2 |
144 |
3350749 |
BC065926.1 |
- |
1, 2 |
122 |
6164621 |
BX648005.1 |
- |
1, 2 |
244 |
|
DA807043.1 |
- |
1, 2 |
264 |
|
M74090.1 |
- |
1, 2 |
145 |
|
View Full Product Details