Gene Transcripts

Gene Symbol
Phf12
Entrez Gene ID
303274
Gene Name
PHD finger protein 12
Gene Aliases
Chromosome Location
Chr. 10: 64815462 - 64862288 on Build Rnor_6.0
UniGene ID
Rn.101513
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013117.2 NP_001013135.2 9 2513
XM_006246960.2 XP_006247022.1 8 2599
XM_008768005.1 XP_008766227.1 8 2599

View Full Product Details