Gene Transcripts

Gene Symbol
Lrrc58
Entrez Gene ID
303919
Gene Name
leucine rich repeat containing 58
Gene Aliases
RGD1307440
Chromosome Location
Chr. 11: 65747235 - 65759558 on Build Rnor_6.0
UniGene ID
Rn.16536
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195558.1 NP_001182487.1 2 556
GenBank mRNA CB547021.1 - 2 269

View Full Product Details