Gene Transcripts

Gene Symbol
Hvcn1
Entrez Gene ID
304485
Gene Name
hydrogen voltage-gated channel 1
Gene Aliases
RGD1310788
Chromosome Location
Chr. 12: 39822044 - 39851089 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006249369.2 XP_006249431.1 6 2009
XM_008769280.1 XP_008767502.1 9 1176

View Full Product Details