Gene Transcripts

Gene Symbol
Coq5
Entrez Gene ID
304542
Gene Name
coenzyme Q5 homolog, methyltransferase (S. cerevisiae)
Gene Aliases
RGD1310857
Chromosome Location
Chr. 12: 47078753 - 47095438 on Build Rnor_6.0
UniGene ID
Rn.143996
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039022.1 NP_001034111.1 6, 7 878
GenBank mRNA BC098719.1 - 6, 7 878

View Full Product Details