Gene Transcripts
- Gene Symbol
- Limch1
- Entrez Gene ID
- 305332
- Gene Name
- LIM and calponin homology domains 1
- Gene Aliases
- RGD1305269
- Chromosome Location
- Chr. 14: 42760363 - 43072843 on Build Rnor_6.0
- UniGene ID
- Rn.232004
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001191678.1 |
NP_001178607.1
|
25, 26 |
3137 |
|
XM_006250945.2 |
XP_006251007.1
|
20, 21 |
3164 |
|
XM_008770143.1 |
XP_008768365.1
|
22, 23 |
3320 |
|
XM_008770144.1 |
XP_008768366.1
|
22, 23 |
3311 |
|
XM_008770145.1 |
XP_008768367.1
|
21, 22 |
3284 |
|
XM_008770146.1 |
XP_008768368.1
|
21, 22 |
3251 |
|
XM_008770147.1 |
XP_008768369.1
|
25, 26 |
4710 |
|
XM_008770148.1 |
XP_008768370.1
|
21, 22 |
3233 |
|
XM_008770149.1 |
XP_008768371.1
|
20, 21 |
3215 |
|
XM_008770150.1 |
XP_008768372.1
|
20, 21 |
3197 |
|
XM_008770151.1 |
XP_008768373.1
|
19, 20 |
3128 |
|
GenBank mRNA |
FQ226534.1 |
- |
13, 14 |
1648 |
|
View Full Product Details