Gene Transcripts

Gene Symbol
Slc35f4
Entrez Gene ID
305865
Gene Name
solute carrier family 35, member F4
Gene Aliases
RGD1561109
Chromosome Location
Chr. 15: 26151800 - 26175678 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008770650.1 XP_008768872.1 6 1151

View Full Product Details