Gene Transcripts

Gene Symbol
Nlrp9
Entrez Gene ID
292577
Gene Name
NLR family, pyrin domain containing 9
Gene Aliases
Nalp9a, Nalp9b, Nlrp9b
Chromosome Location
Chr. 1: 71859390 - 71909138 on Build Rnor_6.0
UniGene ID
Rn.222429
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001169149.1 NP_001162620.1 2 1722
XM_006228188.1 XP_006228250.1 2 1722

View Full Product Details