Gene Transcripts

Gene Symbol
Fnbp1l
Entrez Gene ID
310839
Gene Name
formin binding protein 1-like
Gene Aliases
RGD1305386, Toca-1
Chromosome Location
Chr. 2: 226691435 - 226742081 on Build Rnor_6.0
UniGene ID
Rn.213503
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039609.1 NP_001034698.1 1387
GenBank mRNA AB250295.1 - 1387
AB250296.1 - 1213

View Full Product Details