Gene Transcripts
- Gene Symbol
- Dppa5
- Entrez Gene ID
- 301101
- Gene Name
- developmental pluripotency associated 5
- Gene Aliases
- Chromosome Location
- Chr. 8: 85711560 - 85713316 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_236761.3 |
XP_236761.1
|
2 |
801 |
|
- Gene Symbol
- RGD1559995
- Entrez Gene ID
- 299829
- Gene Name
- similar to developmental pluripotency associated 5
- Gene Aliases
- Chromosome Location
- Chr. 7: 69982196 - 69982644 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_235194.3 |
XP_235194.1
|
1 |
181 |
|
- Gene Symbol
- RGD1564306
- Entrez Gene ID
- 311031
- Gene Name
- similar to developmental pluripotency associated 5
- Gene Aliases
- Chromosome Location
- Chr. 3: 38367482 - 38368105 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_229957.4 |
XP_229957.2
|
1 |
203 |
|
View Full Product Details