Gene Transcripts

Gene Symbol
Hao1
Entrez Gene ID
311446
Gene Name
hydroxyacid oxidase (glycolate oxidase) 1
Gene Aliases
Gox1, XDH1
Chromosome Location
Chr. 3: 127444900 - 127500709 on Build Rnor_6.0
UniGene ID
Rn.126636
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107780.2 NP_001101250.1 5 783
GenBank mRNA BC158804.1 - 5 783

View Full Product Details