Gene Transcripts

Gene Symbol
PRSS12
Entrez Gene ID
8492
Gene Name
protease, serine 12
Gene Aliases
BSSP-3, BSSP3, MRT1
Chromosome Location
Chr. 4: 118280038 - 118353042 on Build GRCh38
UniGene ID
Hs.445857
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003619.3 NP_003610.2 12 2113
GenBank mRNA AF077298.1 - 3 224
AJ001531.1 - 12 2106

View Full Product Details