Gene Transcripts

Gene Symbol
Ns5atp4
Entrez Gene ID
311934
Gene Name
NS5A (hepatitis C virus) transactivated protein 4
Gene Aliases
NICE-3
Chromosome Location
Chr. 3: 35542152 - 35542907 on Build Rnor_6.0
UniGene ID
Rn.124448
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207607.1 NP_997490.1 1 117
GenBank mRNA AY533136.1 - 1 117

View Full Product Details