Gene Transcripts

Gene Symbol
Dcaf10
Entrez Gene ID
313242
Gene Name
DDB1 and CUL4 associated factor 10
Gene Aliases
RGD1304730, Wdr32
Chromosome Location
Chr. 5: 60900414 - 60937547 on Build Rnor_6.0
UniGene ID
Rn.199359
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107935.1 NP_001101405.1 5, 6 1212
XM_006238066.2 XP_006238128.1 4, 5 1050

View Full Product Details