Gene Transcripts

Gene Symbol
Vps13d
Entrez Gene ID
313825
Gene Name
vacuolar protein sorting 13 homolog D (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 5: 162891451 - 163119239 on Build Rnor_6.0
UniGene ID
Rn.2644
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108006.1 NP_001101476.1 19 3683
XM_006239349.2 XP_006239411.1 19 3678
XM_006239350.2 XP_006239412.1 19 3648
XM_006239351.2 XP_006239413.1 19 3645
XM_006239352.2 XP_006239414.1 19 3647
XM_006239353.2 XP_006239415.1 19 3647

View Full Product Details